Assay Details
Gene Symbol: TRAF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190945.1 | NP_001177874.1 | 7 - 8 | 1308 | 89 | |
NM_001190947.1 | NP_001177876.1 | 4 - 5 | 720 | 89 | ||
NM_005658.4 | NP_005649.1 | 6 - 7 | 1455 | 89 | ||
GenBank mRNA | AK090468.1 | - | 6 - 7 | 1456 | 89 | |
AK295959.1 | - | 4 - 5 | 720 | 89 | ||
AK315476.1 | - | 7 - 8 | 1336 | 89 | ||
AL832989.1 | - | 6 - 7 | 3328 | 89 | ||
BC024145.2 | - | 5 - 6 | 961 | 3832475 | 89 | |
BT019408.1 | - | 5 - 6 | 883 | 89 | ||
U19261.1 | - | 5 - 6 | 958 | 89 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09001541.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
signal transduction
regulation of tumor necrosis factor-mediated signaling pathway
positive regulation of NF-kappaB transcription factor activity
protein-containing complex assembly
regulation of extrinsic apoptotic signaling pathway
tumor necrosis factor receptor binding
protein binding
zinc ion binding
ubiquitin protein ligase binding
thioesterase binding
identical protein binding
apoptotic process
signal transduction
regulation of tumor necrosis factor-mediated signaling pathway
positive regulation of NF-kappaB transcription factor activity
protein-containing complex assembly
regulation of extrinsic apoptotic signaling pathway
tumor necrosis factor receptor binding
protein binding
zinc ion binding
ubiquitin protein ligase binding
thioesterase binding
identical protein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09019371.hg.1 | ENST00000373887.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09019371.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09019371.hg.1 | ENST00000546084.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09019371.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09019371.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09019371.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09019371.hg.1 | TC09001541.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR09019370.hg.1 | ENST00000373887.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09019370.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09019370.hg.1 | ENST00000546084.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09019370.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09019370.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09019370.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09019370.hg.1 | TC09001541.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900011393.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900177677.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900177677.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900177677.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900177677.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900177677.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900177677.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900177677.hg.1 | TC0900011393.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900177677.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900177677.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900177677.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900177677.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900177677.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900177677.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900177676.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900177676.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900177676.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900177676.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900177676.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900177676.hg.1 | NM_005658.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900177676.hg.1 | TC0900011393.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900177676.hg.1 | ENST00000373887.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900177676.hg.1 | ENST00000540010.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900177676.hg.1 | ENST00000546084.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900177676.hg.1 | NM_001190945.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900177676.hg.1 | NM_001190947.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900177676.hg.1 | NM_005658.4 | Refseq |
