Assay Details
Gene Symbol: CHRNA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ584708.1 | - | 1 - 2 | 189 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000558903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000558903.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000559941.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000326828.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000348639.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000558903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000558903.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559002.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559080.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559658.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000559941.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | ENST00000561128.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_000743.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NM_001166694.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010151.hg.1 | NR_046313.1 | Refseq |
