Assay Details
Gene Symbol: PI4KB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002651.2 | NP_002642.1 | 2 - 3 | 594 | 80 | |
XM_005245264.3 | XP_005245321.1 | 2 - 3 | 594 | 80 | ||
XM_011509634.1 | XP_011507936.1 | 2 - 3 | 594 | 80 | ||
XM_011509638.1 | XP_011507940.1 | 2 - 3 | 594 | 80 | ||
GenBank mRNA | AB005910.1 | - | 1 - 2 | 82 | 80 | |
AJ011121.1 | - | 1 - 2 | 67 | 80 | ||
AK311292.1 | - | 2 - 3 | 594 | 80 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01003223.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
