Assay Details
Gene Symbol: ATAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024857.4 | NP_079133.3 | 4 - 5 | 2616 | 146 | |
XM_011525269.2 | XP_011523571.1 | 4 - 5 | 2637 | 146 | ||
XM_011525270.2 | XP_011523572.1 | 4 - 5 | 2637 | 146 | ||
XM_011525273.2 | XP_011523575.1 | 4 - 5 | 3654 | 146 | ||
XM_017025111.1 | XP_016880600.1 | 4 - 5 | 2612 | 146 | ||
GenBank mRNA | AJ314648.2 | - | 4 - 5 | 2584 | 146 | |
AK022797.1 | - | 3 - 4 | 1111 | 146 | ||
AL832103.1 | - | 4 - 5 | 2617 | 146 | ||
AY557611.1 | - | 4 - 5 | 2584 | 146 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000355.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17004444.hg.1 | ENST00000321990.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17004444.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17004444.hg.1 | TC17000355.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000390888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000390888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700153471.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153471.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153471.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153471.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700153471.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153471.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153471.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153471.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700153471.hg.1 | TC1700007469.hg.1 | Clariom D TC |
