Assay Details
Gene Symbol: ATAD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024857.4 | NP_079133.3 | 9 - 10 | 3341 | 74 | |
GenBank mRNA | AJ314648.2 | - | 9 - 10 | 3309 | 74 | |
AK022797.1 | - | 8 - 9 | 1836 | 74 | ||
AL832103.1 | - | 9 - 10 | 3342 | 74 | ||
AY557611.1 | - | 9 - 10 | 3309 | 74 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000355.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000390888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17004449.hg.1 | ENST00000321990.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17004449.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17004449.hg.1 | TC17000355.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700153482.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153482.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153482.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153482.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700153482.hg.1 | TC1700007469.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700153482.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700153482.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153482.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700153482.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000321990.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000390888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000578295.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | ENST00000585133.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007469.hg.1 | NM_024857.4 | Refseq |
