Assay Details
Gene Symbol: GAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017017529.1 | XP_016873018.1 | 1 - 2 | 125 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017017529.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000526665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000526665.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000278187.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000454584.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000524701.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000526665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000526665.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000528582.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000532398.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533092.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000533363.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000534801.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | ENST00000630668.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_001143830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_005256.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100007069.hg.1 | NM_177553.2 | Refseq |
