Assay Details
Gene Symbol: NET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047160.2 | NP_001040625.1 | 3 - 4 | 395 | 116 | |
GenBank mRNA | AB209852.1 | - | 3 - 4 | 341 | 116 | |
AK299335.1 | - | 3 - 4 | 351 | 116 | ||
AK302687.1 | - | 3 - 4 | 144 | 116 | ||
BC053553.1 | - | 3 - 4 | 347 | 6137683 | 116 | |
BP354878.1 | - | 3 - 4 | 391 | 116 | ||
DC307907.1 | - | 3 - 4 | 395 | 116 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10000047.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK302687.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
