Assay Details
Gene Symbol: ANK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017016126.1 | XP_016871615.1 | 22 - 23 | 2829 | 117 | |
XM_017016127.1 | XP_016871616.1 | 22 - 23 | 2829 | 117 | ||
XM_017016134.1 | XP_016871623.1 | 22 - 23 | 2829 | 117 | ||
XM_017016141.1 | XP_016871630.1 | 22 - 23 | 2829 | 117 | ||
XM_017016142.1 | XP_016871631.1 | 22 - 23 | 2829 | 117 | ||
XM_017016143.1 | XP_016871632.1 | 22 - 23 | 2829 | 117 | ||
XM_017016144.1 | XP_016871633.1 | 22 - 23 | 2829 | 117 | ||
XM_017016145.1 | XP_016871634.1 | 22 - 23 | 2829 | 117 | ||
XM_017016146.1 | XP_016871635.1 | 22 - 23 | 2829 | 117 | ||
XM_017016147.1 | XP_016871636.1 | 22 - 23 | 2829 | 117 | ||
XM_017016148.1 | XP_016871637.1 | 22 - 23 | 2711 | 117 | ||
XM_017016149.1 | XP_016871638.1 | 22 - 23 | 2928 | 117 | ||
GenBank mRNA | AB209308.1 | - | 22 - 23 | 2917 | 117 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
