Assay Details
Gene Symbol: TSPAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005727.3 | NP_005718.2 | 1 - 2 | 332 | 56 | |
XM_011540460.2 | XP_011538762.1 | 1 - 2 | 306 | 56 | ||
GenBank mRNA | AK313774.1 | - | 1 - 2 | 322 | 56 | |
BM908049.1 | - | 1 - 2 | 335 | 56 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000580.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK313774.1, NM_005727.3, XM_011540460.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01009849.hg.1 | ENST00000372003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009849.hg.1 | ENST00000498443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009849.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01009849.hg.1 | TC01000580.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01009854.hg.1 | ENST00000372003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009854.hg.1 | ENST00000498443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009854.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01009854.hg.1 | TC01000580.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009848.hg.1 | ENST00000372003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009848.hg.1 | ENST00000498443.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009848.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01009848.hg.1 | TC01000580.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158223.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158223.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158223.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158223.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158223.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158223.hg.1 | TC0100008145.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100158223.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158223.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158223.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158223.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158223.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158224.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158224.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158224.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158224.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158224.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158224.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158224.hg.1 | TC0100008145.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100158224.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158224.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158224.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158224.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158224.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158224.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158228.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158228.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158228.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158228.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158228.hg.1 | TC0100008145.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100158228.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158228.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158228.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158228.hg.1 | NM_005727.3 | Refseq |
