Assay Details
Gene Symbol: TSPAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005727.3 | NP_005718.2 | 8 - 9 | 1154 | 68 | |
GenBank mRNA | AF054838.1 | - | 6 - 7 | 680 | 68 | |
AF065388.1 | - | 7 - 8 | 801 | 68 | ||
AK313774.1 | - | 8 - 9 | 1144 | 68 | ||
BC007290.1 | - | 7 - 8 | 807 | 3349295 | 68 | |
BC013404.2 | - | 7 - 8 | 805 | 2959379 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000580.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01009874.hg.1 | ENST00000372003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009874.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01009874.hg.1 | TC01000580.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01009875.hg.1 | ENST00000372003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009875.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01009875.hg.1 | TC01000580.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009872.hg.1 | ENST00000372003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009872.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01009872.hg.1 | TC01000580.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01009873.hg.1 | ENST00000372003.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01009873.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01009873.hg.1 | TC01000580.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0100158249.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158249.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158249.hg.1 | TC0100008145.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100158249.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158249.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158248.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158248.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158248.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158248.hg.1 | TC0100008145.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100158248.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158248.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158248.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000498443.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000464786.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000469330.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000472170.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482143.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008145.hg.1 | ENST00000482928.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158246.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158246.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158246.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158246.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158246.hg.1 | TC0100008145.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100158246.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158246.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158246.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158246.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158247.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158247.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158247.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158247.hg.1 | NM_005727.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100158247.hg.1 | TC0100008145.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100158247.hg.1 | ENST00000372003.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158247.hg.1 | ENST00000470318.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158247.hg.1 | ENST00000475163.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100158247.hg.1 | NM_005727.3 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 106461 | - | - |
