Assay Details
Gene Symbol: PHYH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037537.1 | NP_001032626.1 | 4 - 5 | 687 | 107 | |
NM_001323080.1 | NP_001310009.1 | 5 - 6 | 746 | 107 | ||
NM_006214.3 | NP_006205.1 | 5 - 6 | 582 | 107 | ||
GenBank mRNA | AF023462.1 | - | 5 - 6 | 521 | 107 | |
AF112977.1 | - | 5 - 6 | 553 | 107 | ||
BC029512.1 | - | 5 - 6 | 542 | 5277185 | 107 | |
CR542055.1 | - | 5 - 6 | 494 | 107 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001048.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10012987.hg.1 | ENST00000263038.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10012987.hg.1 | ENST00000396913.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10012987.hg.1 | ENST00000396920.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10012987.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10012987.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10012987.hg.1 | TC10001048.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | TC1000009841.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000167088.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000263038.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396913.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000396920.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000453759.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000463730.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000464049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | ENST00000479604.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_001037537.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009841.hg.1 | NM_006214.3 | Refseq |
