Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006561.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006561.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006561.hg.1 |
TC11000541.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006563.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006563.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006563.hg.1 |
TC11000541.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC0100009017.hg.1 |
ENST00000401028.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000535307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000535307.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000535723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000535723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
ENST00000535723.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006560.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006560.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11006560.hg.1 |
TC11000541.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100200842.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100200842.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100200842.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100200842.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100200842.hg.1 |
TC1100013021.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100200844.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100200844.hg.1 |
ENST00000535723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100200844.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100200844.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100200844.hg.1 |
ENST00000535723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100200844.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100200844.hg.1 |
TC1100013021.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100009017.hg.1 |
ENST00000401028.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100200843.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100200843.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100200843.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100200843.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100200843.hg.1 |
TC1100013021.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100200847.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100200847.hg.1 |
ENST00000535723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100200847.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100200847.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100200847.hg.1 |
ENST00000535723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100200847.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100200847.hg.1 |
TC1100013021.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000305885.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000535307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000535307.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000535723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000535723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
ENST00000535723.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013021.hg.1 |
NM_004111.5 |
Refseq |