Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026543.hg.1 |
TC06002114.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026543.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026543.hg.1 |
ENST00000367941.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026543.hg.1 |
ENST00000448348.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026543.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06026543.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013193.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191094.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191094.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191094.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191094.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191094.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191094.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191094.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191094.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191094.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191094.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191094.hg.1 |
TC0600013193.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0600191093.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191093.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191093.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191093.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191093.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191093.hg.1 |
ENST00000367937.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191093.hg.1 |
ENST00000367941.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191093.hg.1 |
ENST00000448348.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191093.hg.1 |
ENST00000475879.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191093.hg.1 |
NM_003569.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191093.hg.1 |
TC0600013193.hg.1 |
Clariom D TC |