Assay Details
Gene Symbol: BRI3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159491.1 | NP_001152963.1 | 1 | 94 | 140 | |
NM_015379.4 | NP_056194.1 | 1 | 94 | 140 | ||
XM_017011936.1 | XP_016867425.1 | 1 | 88 | 140 | ||
GenBank mRNA | BQ083249.1 | - | 1 | 94 | 140 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC07000584.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001159491.1, NM_015379.4, XM_017011936.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR07008512.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07008512.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07008512.hg.1 | ENST00000539286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07008512.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07008512.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07008512.hg.1 | TC07000584.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1200010148.hg.1 | ENST00000396004.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07008510.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07008510.hg.1 | ENST00000539286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07008510.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07008510.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07008510.hg.1 | TC07000584.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR07008521.hg.1 | ENST00000539286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07008521.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07008521.hg.1 | TC07000584.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0100009208.hg.1 | ENST00000440440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000474291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000474291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000474291.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07008509.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07008509.hg.1 | ENST00000539286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR07008509.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07008509.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR07008509.hg.1 | TC07000584.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0700197755.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197755.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197755.hg.1 | ENST00000474291.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197755.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197755.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197755.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197755.hg.1 | TC0700013405.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700197755.hg.1 | ENST00000474291.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197755.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197755.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197755.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197755.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197755.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197765.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197765.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197765.hg.1 | TC0700013405.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700197765.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197765.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009208.hg.1 | ENST00000440440.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197751.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197751.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197751.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197751.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197751.hg.1 | TC0700013405.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700197751.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197751.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197751.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197751.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197766.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197766.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197766.hg.1 | TC0700013405.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700197766.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197766.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197753.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197753.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197753.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197753.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197753.hg.1 | TC0700013405.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700197753.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197753.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197753.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197753.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197752.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197752.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197752.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197752.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197752.hg.1 | TC0700013405.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700197752.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197752.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700197752.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700197752.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010148.hg.1 | ENST00000396004.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000297290.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000473967.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000474291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000474291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000474291.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | ENST00000539286.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_001159491.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_015379.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013405.hg.1 | NM_015379.4 | Refseq |
