Assay Details
Gene Symbol: CCDC186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001321829.1 | NP_001308758.1 | 15 - 16 | 2784 | 83 | |
NM_018017.3 | NP_060487.2 | 14 - 15 | 2659 | 83 | ||
XM_011539915.2 | XP_011538217.1 | 13 - 14 | 2002 | 83 | ||
GenBank mRNA | AK001050.1 | - | 4 - 5 | 553 | 83 | |
BC100670.1 | - | 15 - 16 | 2711 | 83 | ||
BC103499.1 | - | 14 - 15 | 2586 | 40001826 | 83 | |
BC105632.1 | - | 14 - 15 | 2586 | 40001824 | 83 | |
BC152306.1 | - | 14 - 15 | 2590 | 40012131 | 83 | |
BX537964.1 | - | 14 - 15 | 2631 | 83 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001674.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR10021968.hg.1 | ENST00000369287.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021968.hg.1 | ENST00000430353.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021968.hg.1 | ENST00000497592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021968.hg.1 | ENST00000543782.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021968.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10021968.hg.1 | TC10001674.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10021964.hg.1 | ENST00000369287.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021964.hg.1 | ENST00000430353.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021964.hg.1 | ENST00000543782.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10021964.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10021964.hg.1 | TC10001674.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000459421.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000459421.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000490661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000490661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000490661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000497592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | ENST00000497592.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NR_031747.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000011892.hg.1 | NR_031747.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369285.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000459421.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000459421.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000490661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000490661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000490661.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000497592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | ENST00000497592.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NR_031747.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000011892.hg.1 | NR_031747.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000181387.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181387.hg.1 | ENST00000428953.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181387.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000181387.hg.1 | TC1000011892.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1000181387.hg.1 | NM_018017.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000181387.hg.1 | ENST00000369287.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000181387.hg.1 | ENST00000428953.1 | Ensembl |
