Assay Details
Gene Symbol: BOK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011511696.2 | XP_011509998.1 | 1 - 2 | 64 | 72 | |
XM_017004776.1 | XP_016860265.1 | 1 - 2 | 94 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011511696.2, XM_017004776.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
