Assay Details
Gene Symbol: CBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003655.2 | NP_003646.2 | 4 - 5 | 409 | 57 | |
XM_011525399.2 | XP_011523701.1 | 2 - 3 | 1918 | 57 | ||
GenBank mRNA | AF013956.1 | - | 4 - 5 | 412 | 57 | |
AY390430.1 | - | 4 - 5 | 246 | 57 | ||
BC014967.1 | - | 4 - 5 | 265 | 4856728 | 57 | |
BG029608.1 | - | 4 - 5 | 242 | 57 | ||
BQ225831.1 | - | 409 | 57 | |||
EU439707.1 | - | 4 - 5 | 441 | 57 | ||
KU178422.1 | - | 4 - 5 | 246 | 57 | ||
KU178423.1 | - | 4 - 5 | 246 | 57 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001930.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
transcription, DNA-templated
protein sumoylation
negative regulation of apoptotic process
negative regulation of transcription, DNA-templated
transcription cis-regulatory region binding
chromatin binding
transcription corepressor activity
single-stranded RNA binding
protein binding
ligase activity
SUMO transferase activity
enzyme binding
SUMO binding
methylated histone binding
phosphoprotein binding
negative regulation of transcription by RNA polymerase II
transcription, DNA-templated
protein sumoylation
negative regulation of apoptotic process
negative regulation of transcription, DNA-templated
transcription cis-regulatory region binding
chromatin binding
transcription corepressor activity
single-stranded RNA binding
protein binding
ligase activity
SUMO transferase activity
enzyme binding
SUMO binding
methylated histone binding
phosphoprotein binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17026289.hg.1 | ENST00000269397.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026289.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026289.hg.1 | TC17001930.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17026290.hg.1 | ENST00000269397.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026290.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026290.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026290.hg.1 | TC17001930.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026288.hg.1 | ENST00000269397.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026288.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026288.hg.1 | TC17001930.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17026287.hg.1 | ENST00000269397.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026287.hg.1 | ENST00000343048.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026287.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026287.hg.1 | TC17001930.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17026291.hg.1 | ENST00000269397.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026291.hg.1 | ENST00000343048.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026291.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17026291.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17026291.hg.1 | TC17001930.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700197627.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197627.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197627.hg.1 | TC1700011953.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700197627.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197627.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197630.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197630.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197630.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197630.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197630.hg.1 | TC1700011953.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700197630.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197630.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197630.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197630.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197629.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197629.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197629.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197629.hg.1 | TC1700011953.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700197629.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197629.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197629.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197626.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197626.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197626.hg.1 | TC1700011953.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700197626.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197626.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197631.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197631.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197631.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197631.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197631.hg.1 | TC1700011953.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700197631.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197631.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197631.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197631.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000448310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000494546.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | ENST00000495122.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011953.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197628.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197628.hg.1 | NM_003655.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700197628.hg.1 | TC1700011953.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700197628.hg.1 | ENST00000269397.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700197628.hg.1 | NM_003655.2 | Refseq |
