Assay Details
Gene Symbol: TNFAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021137.4 | NP_066960.1 | 3 - 4 | 825 | 101 | |
XM_017024993.1 | XP_016880482.1 | 3 - 4 | 781 | 101 | ||
GenBank mRNA | AK300584.1 | - | 2 - 3 | 267 | 101 | |
AK312387.1 | - | 3 - 4 | 586 | 101 | ||
BC001643.2 | - | 3 - 4 | 572 | 2823154 | 101 | |
BC001949.1 | - | 3 - 4 | 572 | 2823154 | 101 | |
BC003694.2 | - | 3 - 4 | 587 | 101 | ||
BT020121.1 | - | 2 - 3 | 376 | 101 | ||
CR456819.1 | - | 2 - 3 | 376 | 101 | ||
M80783.1 | - | 3 - 4 | 529 | 101 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000320.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK300584.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication
apoptotic process
immune response
embryo development
cell migration
protein ubiquitination
negative regulation of Rho protein signal transduction
stress fiber assembly
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of DNA replication
protein homooligomerization
ubiquitin-protein transferase activity
protein binding
protein domain specific binding
DNA replication
apoptotic process
immune response
embryo development
cell migration
protein ubiquitination
negative regulation of Rho protein signal transduction
stress fiber assembly
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of DNA replication
protein homooligomerization
ubiquitin-protein transferase activity
protein binding
protein domain specific binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17003847.hg.1 | ENST00000226225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17003847.hg.1 | ENST00000544907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17003847.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17003847.hg.1 | TC17000320.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17003848.hg.1 | ENST00000226225.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17003848.hg.1 | ENST00000544907.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17003848.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17003848.hg.1 | TC17000320.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700152304.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152304.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152304.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152304.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152304.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152304.hg.1 | TC1700007361.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700152304.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152304.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152304.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152304.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152304.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152305.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152305.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152305.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152305.hg.1 | ENST00000583213.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152305.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152305.hg.1 | TC1700007361.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700152305.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152305.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152305.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152305.hg.1 | ENST00000583213.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152305.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152302.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152302.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152302.hg.1 | ENST00000578158.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152302.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152302.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700152302.hg.1 | TC1700007361.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700152302.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152302.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152302.hg.1 | ENST00000578158.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152302.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700152302.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000226225.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000544907.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000577535.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000578158.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000582302.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | ENST00000583213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007361.hg.1 | NM_021137.4 | Refseq |
