Assay Details
Gene Symbol: KRBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205272.1 | NP_001192201.1 | 4 - 5 | 436 | 137 | |
GenBank mRNA | AK130974.1 | - | 4 - 5 | 418 | 137 | |
BC062786.1 | - | 4 - 5 | 422 | 137 | ||
BC103985.1 | - | 3 - 4 | 481 | 137 | ||
BC103986.1 | - | 3 - 4 | 481 | 137 | ||
BC103987.1 | - | 3 - 4 | 481 | 137 | ||
BC118551.1 | - | 3 - 4 | 481 | 137 | ||
BC118662.1 | - | 3 - 4 | 481 | 137 | ||
DB458286.2 | - | 4 - 5 | 418 | 137 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03003346.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02199015 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332717 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02199024 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02199000 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02198997 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02199011 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02199014 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332722 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02198998 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02199022 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02198983 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02199028 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332719 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02198996 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332713 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02199025 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332725 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332738 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332733 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332720 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02199033 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332745 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332716 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332721 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332739 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332731 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02198988 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02199021 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02198987 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02198992 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332724 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02198985 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332744 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02199032 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332741 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02198984 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22332711 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P02199009 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P80551 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 41607 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 56697 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P80551 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P80551 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03004606.hg.1 | ENST00000383748.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03004606.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03004606.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03004606.hg.1 | TC03003346.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0300013815.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300203872.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203872.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203872.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203872.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300203872.hg.1 | TC0300013815.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300203872.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300203872.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203872.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203872.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203873.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203873.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203873.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203873.hg.1 | NM_001205272.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300203873.hg.1 | TC0300013815.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0300203873.hg.1 | ENST00000383748.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203873.hg.1 | ENST00000418176.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203873.hg.1 | ENST00000463888.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300203873.hg.1 | NM_001205272.1 | Refseq |
