Assay Details
Gene Symbol: MAP3K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006609.4 | NP_006600.3 | 7 - 8 | 695 | 108 | |
GenBank mRNA | AB208963.1 | - | 6 - 7 | 601 | 108 | |
AK315007.1 | - | 8 - 9 | 880 | 108 | ||
BC136293.1 | - | 7 - 8 | 664 | 9020280 | 108 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002279.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
positive regulation of transcription, DNA-templated
cellular response to mechanical stimulus
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
protein kinase binding
metal ion binding
protein phosphorylation
positive regulation of transcription, DNA-templated
cellular response to mechanical stimulus
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
protein binding
ATP binding
protein kinase binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02036643.hg.1 | ENST00000344908.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02036643.hg.1 | ENST00000409947.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02036643.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02036643.hg.1 | TC02002279.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409179.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409179.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409179.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409179.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409179.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409179.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0200014170.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200203496.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200203496.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200203496.hg.1 | NM_006609.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200203496.hg.1 | TC0200014170.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200203496.hg.1 | ENST00000344908.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200203496.hg.1 | ENST00000409947.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200203496.hg.1 | NM_006609.4 | Refseq |
