Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000624180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000624180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000624180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
ENST00000624180.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100008473.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010354.hg.1 |
ENST00000198801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010354.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010354.hg.1 |
TC11000811.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010356.hg.1 |
ENST00000198801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010356.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010356.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11010356.hg.1 |
TC11000811.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000624180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000624180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000624180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
ENST00000624180.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100008473.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100162735.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162735.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162735.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100162735.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100162735.hg.1 |
TC1100008473.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100162735.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162735.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162737.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162737.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162737.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162737.hg.1 |
NM_025098.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100162737.hg.1 |
TC1100008473.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100162737.hg.1 |
ENST00000198801.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162737.hg.1 |
ENST00000525093.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162737.hg.1 |
ENST00000526712.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100162737.hg.1 |
NM_025098.3 |
Refseq |