Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000482345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000482345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
ENST00000482345.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005650.hg.1 |
ENST00000319298.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005650.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005650.hg.1 |
ENST00000540661.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005650.hg.1 |
ENST00000541021.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005650.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005650.hg.1 |
TC13000498.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1300156613.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156613.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300156613.hg.1 |
TC1300008386.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1300156613.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300156613.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000381801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000482345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000482345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
ENST00000482345.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008386.hg.1 |
NM_004685.3 |
Refseq |