Assay Details
Gene Symbol: NNMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006169.2 | NP_006160.1 | 1 - 2 | 896 | 143 | |
GenBank mRNA | AK026845.1 | - | 1 - 2 | 272 | 143 | |
BC000234.1 | - | 3 - 4 | 416 | 3350358 | 143 | |
CR541907.1 | - | 1 - 2 | 152 | 143 | ||
U08021.1 | - | 1 - 2 | 269 | 143 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001027.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR11012854.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11012854.hg.1 | ENST00000535401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11012854.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11012854.hg.1 | TC11001027.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR11012853.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11012853.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11012853.hg.1 | TC11001027.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541090.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | ENST00000627588.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11012856.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11012856.hg.1 | ENST00000535401.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11012856.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR11012856.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR11012856.hg.1 | TC11001027.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535185.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541090.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000542647.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000545255.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | ENST00000627588.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1100009110.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100167539.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167539.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100167539.hg.1 | TC1100009110.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100167539.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100167539.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167544.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167544.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167544.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167544.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167544.hg.1 | NM_006169.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1100167544.hg.1 | TC1100009110.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1100167544.hg.1 | ENST00000299964.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167544.hg.1 | ENST00000535401.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167544.hg.1 | ENST00000541754.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167544.hg.1 | ENST00000546313.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1100167544.hg.1 | NM_006169.2 | Refseq |
