Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033604.hg.1 |
TC03001862.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033604.hg.1 |
ENST00000316549.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033604.hg.1 |
ENST00000450105.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03033604.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000474151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000474151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000474151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000474151.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000474727.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000474727.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000474727.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000474727.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000474727.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000483124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000483124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000483124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000483124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000483124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000483124.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000498717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000498717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
ENST00000498717.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194828.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194828.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194828.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300194828.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300194828.hg.1 |
TC0300012686.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000316549.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000474151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000474151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000474151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000474151.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000474727.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000474727.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000474727.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000474727.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000474727.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000483124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000483124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000483124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000483124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000483124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000483124.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000498717.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000498717.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
ENST00000498717.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012686.hg.1 |
NM_173653.3 |
Refseq |