Assay Details
Gene Symbol: SLC27A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_003645.3 | NP_003636.2 | 2 - 3 | 914 | 107 | |
GenBank mRNA | AF096290.1 | - | 2 - 3 | 886 | 107 | |
AK222859.1 | - | 2 - 3 | 885 | 107 | ||
AK223145.1 | - | 2 - 3 | 883 | 107 | ||
AK290262.1 | - | 2 - 3 | 914 | 107 | ||
D88308.1 | - | 2 - 3 | 904 | 107 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15000391.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid alpha-oxidation
long-chain fatty acid metabolic process
fatty acid beta-oxidation
bile acid biosynthetic process
fatty acid transport
very long-chain fatty acid catabolic process
long-chain fatty acid import
methyl-branched fatty acid metabolic process
long-chain fatty acid-CoA ligase activity
receptor binding
ATP binding
fatty acid transporter activity
enzyme binding
very long-chain fatty acid-CoA ligase activity
phytanate-CoA ligase activity
pristanate-CoA ligase activity
decanoate--CoA ligase activity
fatty acid alpha-oxidation
long-chain fatty acid metabolic process
fatty acid beta-oxidation
bile acid biosynthetic process
fatty acid transport
very long-chain fatty acid catabolic process
long-chain fatty acid import
methyl-branched fatty acid metabolic process
long-chain fatty acid-CoA ligase activity
receptor binding
ATP binding
fatty acid transporter activity
enzyme binding
very long-chain fatty acid-CoA ligase activity
phytanate-CoA ligase activity
pristanate-CoA ligase activity
decanoate--CoA ligase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR15003642.hg.1 | ENST00000267842.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15003642.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15003642.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15003642.hg.1 | TC15000391.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500151306.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500151306.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500151306.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500151306.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500151306.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500151306.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500151306.hg.1 | TC1500007196.hg.1 | Clariom D TC |
