Assay Details
Gene Symbol: SLC27A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159629.1 | NP_001153101.1 | 3 - 4 | 1042 | 81 | |
NM_003645.3 | NP_003636.2 | 4 - 5 | 1201 | 81 | ||
GenBank mRNA | AF096290.1 | - | 4 - 5 | 1173 | 81 | |
AK123905.1 | - | 5 - 6 | 843 | 81 | ||
AK222859.1 | - | 4 - 5 | 1172 | 81 | ||
AK223145.1 | - | 4 - 5 | 1170 | 81 | ||
AK290262.1 | - | 4 - 5 | 1201 | 81 | ||
BC057770.1 | - | 3 - 4 | 1005 | 30348317 | 81 | |
D88308.1 | - | 4 - 5 | 1191 | 81 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15000391.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
fatty acid alpha-oxidation
long-chain fatty acid metabolic process
fatty acid beta-oxidation
bile acid biosynthetic process
fatty acid transport
very long-chain fatty acid catabolic process
long-chain fatty acid import into cell
methyl-branched fatty acid metabolic process
long-chain fatty acid-CoA ligase activity
signaling receptor binding
ATP binding
fatty acid transmembrane transporter activity
enzyme binding
very long-chain fatty acid-CoA ligase activity
phytanate-CoA ligase activity
pristanate-CoA ligase activity
decanoate-CoA ligase activity
fatty acid alpha-oxidation
long-chain fatty acid metabolic process
fatty acid beta-oxidation
bile acid biosynthetic process
fatty acid transport
very long-chain fatty acid catabolic process
long-chain fatty acid import into cell
methyl-branched fatty acid metabolic process
long-chain fatty acid-CoA ligase activity
signaling receptor binding
ATP binding
fatty acid transmembrane transporter activity
enzyme binding
very long-chain fatty acid-CoA ligase activity
phytanate-CoA ligase activity
pristanate-CoA ligase activity
decanoate-CoA ligase activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR15003644.hg.1 | ENST00000267842.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15003644.hg.1 | ENST00000380902.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15003644.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15003644.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15003644.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15003644.hg.1 | TC15000391.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500151309.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500151309.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500151309.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500151309.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500151309.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500151309.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500151309.hg.1 | TC1500007196.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500151309.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500151309.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500151309.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500151309.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500151309.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500151309.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000267842.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000380902.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000544960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | ENST00000559938.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_001159629.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500007196.hg.1 | NM_003645.3 | Refseq |
