Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002021.hg.1 |
TC01000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002021.hg.1 |
ENST00000263934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002021.hg.1 |
ENST00000355249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002021.hg.1 |
ENST00000377081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002021.hg.1 |
ENST00000377086.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002021.hg.1 |
NM_015074.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002020.hg.1 |
TC01000127.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002020.hg.1 |
ENST00000263934.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002020.hg.1 |
ENST00000355249.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002020.hg.1 |
ENST00000377081.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002020.hg.1 |
ENST00000377086.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01002020.hg.1 |
NM_015074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147214.hg.1 |
ENST00000263934.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147214.hg.1 |
ENST00000377081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147214.hg.1 |
ENST00000377086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147214.hg.1 |
ENST00000620295.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147214.hg.1 |
ENST00000622724.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147214.hg.1 |
NM_015074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147214.hg.1 |
ENST00000263934.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147214.hg.1 |
ENST00000377081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147214.hg.1 |
ENST00000377086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147214.hg.1 |
ENST00000620295.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147214.hg.1 |
ENST00000622724.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147214.hg.1 |
NM_015074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147214.hg.1 |
TC0100006812.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100147213.hg.1 |
ENST00000263934.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147213.hg.1 |
ENST00000377081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147213.hg.1 |
ENST00000377086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147213.hg.1 |
ENST00000620295.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147213.hg.1 |
ENST00000622724.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147213.hg.1 |
NM_015074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147213.hg.1 |
ENST00000263934.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147213.hg.1 |
ENST00000377081.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147213.hg.1 |
ENST00000377086.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147213.hg.1 |
ENST00000620295.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147213.hg.1 |
ENST00000622724.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100147213.hg.1 |
NM_015074.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100147213.hg.1 |
TC0100006812.hg.1 |
Clariom D TC |