Assay Details
Gene Symbol: GAS2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139285.3 | NP_644814.1 | 3 - 4 | 764 | 70 | |
GenBank mRNA | AF508784.1 | - | 4 - 5 | 717 | 70 | |
AF508785.1 | - | 3 - 4 | 764 | 70 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001375.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17018224.hg.1 | ENST00000254466.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17018224.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17018224.hg.1 | TC17001375.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17018225.hg.1 | ENST00000254466.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17018225.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17018225.hg.1 | TC17001375.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1700181895.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700181895.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700181895.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700181895.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700181895.hg.1 | TC1700010439.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700181895.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700181895.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700181895.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700181895.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604063.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000604641.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | ENST00000618498.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | NM_139285.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700010439.hg.1 | NM_139285.3 | Refseq |
