Assay Details
Gene Symbol: PTPDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253830.1 | NP_001240759.1 | 2 - 3 | 403 | 68 | |
NM_177995.2 | NP_818931.1 | 2 - 3 | 418 | 68 | ||
XM_005251711.4 | XP_005251768.1 | 2 - 3 | 2360 | 68 | ||
XM_011518240.2 | XP_011516542.1 | 2 - 3 | 168 | 68 | ||
XM_011518241.2 | XP_011516543.1 | 2 - 3 | 122 | 68 | ||
XM_017014280.1 | XP_016869769.1 | 3 - 4 | 227 | 68 | ||
XM_017014281.1 | XP_016869770.1 | 2 - 3 | 146 | 68 | ||
GenBank mRNA | BC067120.1 | - | 2 - 3 | 403 | 5539182 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000455.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1100012240.hg.1 | ENST00000533731.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300006613.hg.1 | ENST00000414485.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09004897.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09004897.hg.1 | ENST00000375360.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09004897.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09004897.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09004897.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09004897.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09004897.hg.1 | TC09000455.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC0200009318.hg.1 | ENST00000455007.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600009794.hg.1 | ENST00000565821.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1300008220.hg.1 | ENST00000440548.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010276.hg.1 | ENST00000437729.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010227.hg.1 | ENST00000436426.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008011.hg.1 | ENST00000445119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300008220.hg.1 | ENST00000440548.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | TC0900008010.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153939.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1600009794.hg.1 | ENST00000565821.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008011.hg.1 | ENST00000445119.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010227.hg.1 | ENST00000436426.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1300006613.hg.1 | ENST00000414485.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009318.hg.1 | ENST00000455007.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010276.hg.1 | ENST00000437729.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100012240.hg.1 | ENST00000533731.1 | Ensembl |
