Assay Details
Gene Symbol: PTPDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001253829.1 | NP_001240758.1 | 5 - 6 | 852 | 116 | |
NM_001253830.1 | NP_001240759.1 | 6 - 7 | 913 | 116 | ||
NM_152422.4 | NP_689635.3 | 5 - 6 | 846 | 116 | ||
NM_177995.2 | NP_818931.1 | 6 - 7 | 928 | 116 | ||
XM_005251711.4 | XP_005251768.1 | 6 - 7 | 2870 | 116 | ||
XM_011518240.2 | XP_011516542.1 | 6 - 7 | 678 | 116 | ||
XM_011518241.2 | XP_011516543.1 | 6 - 7 | 632 | 116 | ||
XM_017014280.1 | XP_016869769.1 | 7 - 8 | 737 | 116 | ||
XM_017014281.1 | XP_016869770.1 | 6 - 7 | 656 | 116 | ||
XM_017014282.1 | XP_016869771.1 | 3 - 4 | 759 | 116 | ||
GenBank mRNA | AK124912.1 | - | 5 - 6 | 732 | 116 | |
AK310403.1 | - | 5 - 6 | 810 | 116 | ||
AY171233.1 | - | 6 - 7 | 933 | 116 | ||
BC067120.1 | - | 6 - 7 | 913 | 5539182 | 116 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC09000455.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09004901.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09004901.hg.1 | ENST00000375360.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR09004901.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09004901.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09004901.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09004901.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR09004901.hg.1 | TC09000455.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | TC0900008010.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0900153943.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000288976.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000375360.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000467049.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | ENST00000620992.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253829.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_001253830.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_152422.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008010.hg.1 | NM_177995.2 | Refseq |
