Assay Details
Gene Symbol: GABRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270873.1 | NP_001257802.1 | 3 - 4 | 531 | 74 | |
NM_033223.4 | NP_150092.2 | 3 - 4 | 531 | 74 | ||
XM_011521430.2 | XP_011519732.1 | 3 - 4 | 525 | 74 | ||
GenBank mRNA | BX282033.1 | - | 3 - 4 | 443 | 73 | |
S82769.1 | - | 3 - 4 | 309 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
