Assay Details
Gene Symbol: GABRG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270873.1 | NP_001257802.1 | 4 - 5 | 747 | 103 | |
NM_033223.4 | NP_150092.2 | 4 - 5 | 747 | 103 | ||
XM_011521430.2 | XP_011519732.1 | 4 - 5 | 741 | 103 | ||
XM_017022058.1 | XP_016877547.1 | 2 - 3 | 3232 | 103 | ||
XM_017022059.1 | XP_016877548.1 | 2 - 3 | 572 | 103 | ||
GenBank mRNA | S82769.1 | - | 4 - 5 | 525 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15000125.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000554786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000557596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000557596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15000804.hg.1 | ENST00000333743.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15000804.hg.1 | ENST00000555083.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15000804.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15000804.hg.1 | TC15000125.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000451330.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000553440.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000554786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000557596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000557596.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500006645.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | ENST00000557596.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | NM_033223.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | TC1500006645.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | ENST00000333743.10 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | ENST00000554696.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | ENST00000555083.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | ENST00000557596.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | ENST00000615808.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | NM_001270873.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500145979.hg.1 | NM_033223.4 | Refseq |
