Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009577.hg.1 |
ENST00000437464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009577.hg.1 |
NM_001127464.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009577.hg.1 |
TC16000678.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008821.hg.1 |
ENST00000437464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008821.hg.1 |
ENST00000437464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008821.hg.1 |
ENST00000437464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008821.hg.1 |
ENST00000565624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008821.hg.1 |
ENST00000565624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008821.hg.1 |
ENST00000565624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008821.hg.1 |
ENST00000565624.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008821.hg.1 |
NM_001127464.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008821.hg.1 |
NM_001127464.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600008821.hg.1 |
NM_001127464.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009576.hg.1 |
ENST00000437464.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009576.hg.1 |
NM_001127464.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009576.hg.1 |
TC16000678.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600008821.hg.1 |
ENST00000437464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008821.hg.1 |
ENST00000437464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008821.hg.1 |
ENST00000437464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008821.hg.1 |
ENST00000565624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008821.hg.1 |
ENST00000565624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008821.hg.1 |
ENST00000565624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008821.hg.1 |
ENST00000565624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600008821.hg.1 |
NM_001127464.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008821.hg.1 |
NM_001127464.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600008821.hg.1 |
NM_001127464.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600165653.hg.1 |
ENST00000437464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165653.hg.1 |
ENST00000565624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165653.hg.1 |
NM_001127464.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600165653.hg.1 |
TC1600008821.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600165653.hg.1 |
NM_001127464.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600165653.hg.1 |
ENST00000437464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165653.hg.1 |
ENST00000565624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165651.hg.1 |
ENST00000437464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165651.hg.1 |
ENST00000565624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165651.hg.1 |
NM_001127464.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600165651.hg.1 |
TC1600008821.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600165651.hg.1 |
ENST00000437464.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165651.hg.1 |
ENST00000565624.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600165651.hg.1 |
NM_001127464.2 |
Refseq |