Assay Details
Gene Symbol: CENPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006733.3 | NP_006724.2 | 19 - 20 | 2365 | 79 | |
XM_005262111.2 | XP_005262168.1 | 20 - 21 | 2235 | 79 | ||
XM_011530894.2 | XP_011529196.1 | 20 - 21 | 2264 | 79 | ||
XM_011530895.2 | XP_011529197.1 | 21 - 22 | 2551 | 79 | ||
XM_011530897.2 | XP_011529199.1 | 21 - 22 | 2426 | 79 | ||
XM_011530899.2 | XP_011529201.1 | 19 - 20 | 2232 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000475.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X006149.hg.1 | TC0X000475.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006149.hg.1 | ENST00000372927.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006149.hg.1 | ENST00000423383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006149.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00153905.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153905.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153905.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00153905.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153905.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153905.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00153905.hg.1 | TC0X00007936.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
