Assay Details
Gene Symbol: CENPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318521.1 | NP_001305450.1 | 7 - 8 | 820 | 90 | |
NM_001318523.1 | NP_001305452.1 | 5 - 6 | 657 | 90 | ||
NM_006733.3 | NP_006724.2 | 6 - 7 | 914 | 90 | ||
XM_005262111.2 | XP_005262168.1 | 7 - 8 | 784 | 90 | ||
XM_011530894.2 | XP_011529196.1 | 7 - 8 | 813 | 90 | ||
XM_011530895.2 | XP_011529197.1 | 8 - 9 | 1100 | 90 | ||
XM_011530897.2 | XP_011529199.1 | 8 - 9 | 975 | 90 | ||
XM_011530899.2 | XP_011529201.1 | 6 - 7 | 781 | 90 | ||
XM_017029383.1 | XP_016884872.1 | 7 - 8 | 813 | 90 | ||
GenBank mRNA | AK302986.1 | - | 7 - 8 | 813 | 90 | |
BC012462.2 | - | 5 - 6 | 658 | 4537558 | 90 | |
X97249.1 | - | 6 - 7 | 913 | 90 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000475.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR0X006134.hg.1 | TC0X000475.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006134.hg.1 | ENST00000218507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006134.hg.1 | ENST00000372926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006134.hg.1 | ENST00000372927.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006134.hg.1 | ENST00000423383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006134.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00153878.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153878.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153878.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153878.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00153878.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153878.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153878.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153878.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00153878.hg.1 | TC0X00007936.hg.1 | Clariom D TC |
