Assay Details
Gene Symbol: CENPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001318521.1 | NP_001305450.1 | 8 - 9 | 864 | 68 | |
NM_001318523.1 | NP_001305452.1 | 6 - 7 | 701 | 68 | ||
NM_006733.3 | NP_006724.2 | 7 - 8 | 958 | 68 | ||
XM_005262111.2 | XP_005262168.1 | 8 - 9 | 828 | 68 | ||
XM_011530894.2 | XP_011529196.1 | 8 - 9 | 857 | 68 | ||
XM_011530895.2 | XP_011529197.1 | 9 - 10 | 1144 | 68 | ||
XM_011530897.2 | XP_011529199.1 | 9 - 10 | 1019 | 68 | ||
XM_011530899.2 | XP_011529201.1 | 7 - 8 | 825 | 68 | ||
XM_017029383.1 | XP_016884872.1 | 8 - 9 | 857 | 68 | ||
GenBank mRNA | AK302986.1 | - | 8 - 9 | 857 | 68 | |
BC012462.2 | - | 6 - 7 | 702 | 4537558 | 68 | |
X97249.1 | - | 7 - 8 | 957 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC0X000475.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006135.hg.1 | TC0X000475.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006135.hg.1 | ENST00000218507.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006135.hg.1 | ENST00000372926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006135.hg.1 | ENST00000372927.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006135.hg.1 | ENST00000423383.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR0X006135.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00153880.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153880.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153880.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153880.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00153880.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153880.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153880.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00153880.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00153880.hg.1 | TC0X00007936.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372926.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000372927.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000403304.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000423383.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | ENST00000435570.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00007936.hg.1 | NM_006733.3 | Refseq |
