Assay Details
Gene Symbol: KIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB017133.1 | - | 47 - 48 | 5493 | 90 | |
AB088210.1 | - | 47 - 48 | 5409 | 90 | ||
AB088211.1 | - | 46 - 47 | 5367 | 90 | ||
AB088212.1 | - | 47 - 48 | 5331 | 90 | ||
AB088213.1 | - | 46 - 47 | 5289 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
