Assay Details
Gene Symbol: CDC42EP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006724108.1 | XP_006724171.1 | 1 - 2 | 125 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006724108.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
Rho protein signal transduction
regulation of cell shape
positive regulation of actin filament polymerization
positive regulation of pseudopodium assembly
positive regulation of GTPase activity
cell-cell adhesion
GTPase activator activity
protein binding
cadherin binding involved in cell-cell adhesion
Rho protein signal transduction
regulation of cell shape
positive regulation of actin filament polymerization
positive regulation of pseudopodium assembly
positive regulation of GTPase activity
cell-cell adhesion
GTPase activator activity
protein binding
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000415670.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000415670.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000415670.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000415670.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000430687.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000430687.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000430687.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000430687.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000430687.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000430687.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000434728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000434728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | ENST00000434728.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000249014.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000415670.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000415670.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000415670.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000415670.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000430687.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000430687.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000430687.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000430687.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000430687.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000430687.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000434728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000434728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | ENST00000434728.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | NM_152243.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007303.hg.1 | NM_152243.2 | Refseq |
