Assay Details
Gene Symbol: RGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297669.1 | NP_001284598.1 | 13 - 14 | 1937 | 80 | |
NM_001297670.1 | NP_001284599.1 | 12 - 13 | 1827 | 80 | ||
NM_001297671.1 | NP_001284600.1 | 12 - 13 | 1563 | 80 | ||
NM_015149.4 | NP_055964.3 | 13 - 14 | 1938 | 80 | ||
XM_011509339.2 | XP_011507641.1 | 14 - 15 | 1984 | 80 | ||
XM_011509341.1 | XP_011507643.1 | 13 - 14 | 1891 | 80 | ||
XM_011509342.2 | XP_011507644.1 | 13 - 14 | 1891 | 80 | ||
XM_017000756.1 | XP_016856245.1 | 12 - 13 | 1412 | 80 | ||
GenBank mRNA | AB023176.1 | - | 12 - 13 | 1503 | 80 | |
AF186779.1 | - | 12 - 13 | 1524 | 80 | ||
AF186780.1 | - | 13 - 14 | 1900 | 80 | ||
AK295179.1 | - | 12 - 13 | 1701 | 80 | ||
AK295611.1 | - | 9 - 10 | 1140 | 80 | ||
AK316526.1 | - | 13 - 14 | 1875 | 80 | ||
AL080117.1 | - | 11 - 12 | 1306 | 80 | ||
BC136591.1 | - | 13 - 14 | 1642 | 9020581 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
