Assay Details
Gene Symbol: RGL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297669.1 | NP_001284598.1 | 3 - 4 | 727 | 127 | |
NM_001297670.1 | NP_001284599.1 | 2 - 3 | 617 | 127 | ||
GenBank mRNA | AK295179.1 | - | 2 - 3 | 491 | 127 | |
AK316526.1 | - | 3 - 4 | 665 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
