Assay Details
Gene Symbol: PTTG3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002734.2 | 1 | 464 | 75 | ||
GenBank mRNA | AF095289.1 | - | 1 | 435 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | C8orf44-SGK3 readthrough | C8orf44-SGK3 | Anti-sense | Chr. 8 - 66667552 - 66862022 on NCBI Build GRCh38 | ||
Downstream | minichromosome maintenance domain containing 2 | MCMDC2 | C8orf45 | 102.7kb | Anti-sense | Chr. 8 - 66870749 - 66926436 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P60016 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P60016 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 12255 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 33071 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P60016 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P60016 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26467420 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06184379 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16102576 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16102578 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26467417 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26467418 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16102579 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16102578 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26467417 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P06184379 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16102579 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26467418 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P26467420 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16102576 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P60016 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P60016 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16102587 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184400 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184397 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184410 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16102590 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184414 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184377 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16102588 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184402 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184408 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184386 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184392 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184393 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184383 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184380 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184399 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P06184390 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NR_002734_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 8151124 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8109639 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8151123 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 208511_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11753582_x_at | - | - |
Affymetrix Human Genome U219 Array | 11753582_PM_x_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0400007247.hg.1 | ENST00000504686.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007247.hg.1 | ENST00000504686.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007247.hg.1 | NM_006607.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007247.hg.1 | ENST00000504686.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007247.hg.1 | ENST00000504686.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007247.hg.1 | NM_006607.2 | Refseq |
Human Genome U133 Plus 2.0 Array | 208511_at | - | - |
Human Genome U133 Plus 2.0 Array | 208511_PM_at | - | - |
Human Genome U133A 2.0 Array | 208511_at | - | - |
Human Genome U133A 2.0 Array | 208511_PM_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NR_002734_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw552459 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208511_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208511_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208511_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208511_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208511_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208511_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208511_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208511_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208511_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208511_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 208511_at8 | - | - |
Human Genome U133 Set | 208511_PM_at | - | - |
Human Genome U133 Set | 208511_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035831 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035831 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 147452 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_2049021 | NR_002734.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_2049021 | AF095289.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2049021 | NR_002734.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_2049021 | AF095289.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2049021 | NR_002734.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2049021 | AF095289.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2049021 | NR_002734.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2049021 | AF095289.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2049021 | NR_002734.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2049021 | AF095289.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2049021 | NR_002734.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2049021 | AF095289.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2049021 | NR_002734.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2049021 | AF095289.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2049021 | NR_002734.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2049021 | AF095289.1 | human_transcriptome |
