Assay Details
Gene Symbol: AP5M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018229.3 | NP_060699.3 | 2 - 3 | 1145 | 86 | |
XM_006720196.2 | XP_006720259.1 | 2 - 3 | 774 | 86 | ||
XM_011536940.2 | XP_011535242.1 | 3 - 4 | 1117 | 86 | ||
GenBank mRNA | AF094583.1 | - | 1 - 2 | 110 | 86 | |
AK001675.1 | - | 2 - 3 | 990 | 86 | ||
AK172824.1 | - | 1 - 2 | 207 | 86 | ||
AK225345.1 | - | 2 - 3 | 992 | 86 | ||
AK303999.1 | - | 3 - 4 | 1019 | 86 | ||
AL136685.1 | - | 2 - 3 | 1124 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1100007335.hg.1 | ENST00000604322.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1100007335.hg.1 | ENST00000604322.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150456.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150456.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150456.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150456.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150456.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150456.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150456.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150456.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150456.hg.1 | TC1400007282.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
