Assay Details
Gene Symbol: AP5M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018229.3 | NP_060699.3 | 4 - 5 | 1520 | 135 | |
NR_026895.1 | 3 - 4 | 874 | 135 | |||
XM_006720196.2 | XP_006720259.1 | 4 - 5 | 1149 | 135 | ||
XM_011536940.2 | XP_011535242.1 | 5 - 6 | 1492 | 135 | ||
GenBank mRNA | AK001675.1 | - | 4 - 5 | 1365 | 135 | |
AK172824.1 | - | 3 - 4 | 582 | 135 | ||
AK225345.1 | - | 4 - 5 | 1367 | 135 | ||
AK303999.1 | - | 5 - 6 | 1394 | 135 | ||
AL136685.1 | - | 4 - 5 | 1499 | 135 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14000340.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14003637.hg.1 | ENST00000261558.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003637.hg.1 | ENST00000431972.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003637.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003637.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14003637.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14003637.hg.1 | TC14000340.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150462.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150462.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150462.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150462.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150462.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150462.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150462.hg.1 | TC1400007282.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400150462.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150462.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150462.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150462.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150462.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150462.hg.1 | NR_026895.1 | Refseq |
