Assay Details
Gene Symbol: AP5M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018229.3 | NP_060699.3 | 6 - 7 | 1718 | 97 | |
NR_026895.1 | 5 - 6 | 1072 | 97 | |||
XM_006720196.2 | XP_006720259.1 | 6 - 7 | 1347 | 97 | ||
XM_011536940.2 | XP_011535242.1 | 7 - 8 | 1690 | 97 | ||
GenBank mRNA | AK001675.1 | - | 6 - 7 | 1563 | 97 | |
AK172824.1 | - | 5 - 6 | 780 | 97 | ||
AK225345.1 | - | 6 - 7 | 1565 | 97 | ||
AK303999.1 | - | 7 - 8 | 1592 | 97 | ||
AL136685.1 | - | 6 - 7 | 1697 | 97 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14000340.hg.1
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14003640.hg.1 | ENST00000261558.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003640.hg.1 | ENST00000431972.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14003640.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14003640.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14003640.hg.1 | TC14000340.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1400150466.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150466.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150466.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150466.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150466.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150466.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150466.hg.1 | TC1400007282.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400150466.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150466.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150466.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150466.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400150466.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400150466.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000261558.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000431972.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554213.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554863.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000554931.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000555448.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556377.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556723.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | ENST00000556995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NM_018229.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400007282.hg.1 | NR_026895.1 | Refseq |
