Assay Details
Gene Symbol: IQCA1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001304419.1 | NP_001291348.1 | 9 - 10 | 1247 | 119 | |
GenBank mRNA | HM370405.1 | - | 2 - 3 | 174 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3313352 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700194234.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700194234.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700194234.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700194234.hg.1 | TC0700013064.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700194234.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700194234.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700194234.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700194235.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700194235.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700194235.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0700194235.hg.1 | TC0700013064.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0700194235.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700194235.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0700194235.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000611102.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000613681.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | ENST00000615129.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0700013064.hg.1 | NM_001304419.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1688763 | - | - |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1760518 | - | - |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1690856 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1690856 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1760518 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1669713 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1690856 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1689634 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1689634 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1690856 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1669713 | - | - |
