Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017895.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017895.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017895.hg.1 |
TC10001413.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017894.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017894.hg.1 |
ENST00000466008.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017894.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10017894.hg.1 |
TC10001413.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000175184.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175184.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175184.hg.1 |
TC1000011028.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000175184.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175184.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000011028.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175182.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175182.hg.1 |
NM_021245.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000175182.hg.1 |
TC1000011028.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000175182.hg.1 |
ENST00000359322.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000175182.hg.1 |
NM_021245.3 |
Refseq |