Assay Details
Gene Symbol: GABRA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_000809.3 | NP_000800.2 | 3 - 4 | 1253 | 65 | |
NM_001204266.1 | NP_001191195.1 | 3 - 4 | 283 | 65 | ||
NM_001204267.1 | NP_001191196.1 | 3 - 4 | 283 | 65 | ||
GenBank mRNA | AK090780.1 | - | 3 - 4 | 1253 | 65 | |
BC035055.1 | - | 3 - 4 | 382 | 5259426 | 65 | |
BI599812.1 | - | 3 - 4 | 413 | 65 | ||
CD013892.1 | - | 3 - 4 | 287 | 65 | ||
CD013893.1 | - | 3 - 4 | 287 | 65 | ||
KJ534840.1 | - | 3 - 4 | 270 | 65 | ||
KJ535037.1 | - | 3 - 4 | 270 | 65 | ||
U30461.1 | - | 3 - 4 | 284 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04001165.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transport
gamma-aminobutyric acid signaling pathway
central nervous system development
ion transmembrane transport
chloride transmembrane transport
regulation of response to drug
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
benzodiazepine receptor activity
transport
gamma-aminobutyric acid signaling pathway
central nervous system development
ion transmembrane transport
chloride transmembrane transport
regulation of response to drug
GABA-A receptor activity
extracellular ligand-gated ion channel activity
chloride channel activity
benzodiazepine receptor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015987.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04015987.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015987.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015987.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR04015987.hg.1 | TC04001165.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000509316.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0400010602.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | NM_001204267.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | TC0400010602.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | ENST00000502874.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | ENST00000264318.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | ENST00000508560.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | ENST00000511523.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | NM_000809.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | NM_001204266.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400172251.hg.1 | NM_001204267.1 | Refseq |
