Assay Details
Gene Symbol: DOPEY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320714.1 | NP_001307643.1 | 20 - 21 | 5000 | 132 | |
NM_005128.3 | NP_005119.2 | 20 - 21 | 4891 | 132 | ||
XM_017028509.1 | XP_016883998.1 | 17 - 18 | 4394 | 132 | ||
GenBank mRNA | AB023150.2 | - | 17 - 18 | 4360 | 132 | |
AJ237839.2 | - | 20 - 21 | 4897 | 132 | ||
AK025095.1 | - | 2 - 3 | 492 | 132 | ||
BC012791.2 | - | 2 - 3 | 533 | 132 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC21001061.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR21001278.hg.1 | TC21001061.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR21001278.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR21001278.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000270190.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | ENST00000492760.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007039.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100147524.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100147524.hg.1 | ENST00000463668.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100147524.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100147524.hg.1 | TC2100007039.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100147524.hg.1 | NM_005128.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100147524.hg.1 | ENST00000399151.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100147524.hg.1 | ENST00000463668.1 | Ensembl |
