Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001561.hg.1 |
ENST00000301738.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001561.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16001561.hg.1 |
TC16000082.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000564195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000564195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000564195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000564195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000564195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000564195.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000564246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000564246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000564246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000564246.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000569689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000569689.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000570005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000570005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
ENST00000570005.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146893.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146893.hg.1 |
ENST00000564246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146893.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146893.hg.1 |
TC1600006628.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600146893.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600146893.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600146893.hg.1 |
ENST00000564246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000301738.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000564195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000564195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000564195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000564195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000564195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000564195.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000564246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000564246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000564246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000564246.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000569689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000569689.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000570005.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000570005.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
ENST00000570005.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600006628.hg.1 |
NM_018992.3 |
Refseq |