Assay Details
Gene Symbol: LEO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001323903.1 | NP_001310832.1 | 5 - 6 | 1214 | 106 | |
NM_001323904.1 | NP_001310833.1 | 5 - 6 | 1214 | 106 | ||
NM_138792.3 | NP_620147.1 | 5 - 6 | 1214 | 106 | ||
XM_017021911.1 | XP_016877400.1 | 5 - 6 | 1211 | 106 | ||
GenBank mRNA | AK297972.1 | - | 6 - 7 | 1124 | 106 | |
AY302186.1 | - | 5 - 6 | 1198 | 106 | ||
BC018147.2 | - | 5 - 6 | 1174 | 3847809 | 106 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15001449.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
endodermal cell fate commitment
transcription elongation from RNA polymerase II promoter
mRNA polyadenylation
histone monoubiquitination
Wnt signaling pathway
stem cell population maintenance
positive regulation of mRNA 3'-end processing
positive regulation of transcription elongation from RNA polymerase II promoter
histone H2B ubiquitination
negative regulation of myeloid cell differentiation
positive regulation of transcription by RNA polymerase II
beta-catenin-TCF complex assembly
protein binding
RNA polymerase II C-terminal domain phosphoserine binding
endodermal cell fate commitment
transcription elongation from RNA polymerase II promoter
mRNA polyadenylation
histone monoubiquitination
Wnt signaling pathway
stem cell population maintenance
positive regulation of mRNA 3'-end processing
positive regulation of transcription elongation from RNA polymerase II promoter
histone H2B ubiquitination
negative regulation of myeloid cell differentiation
positive regulation of transcription by RNA polymerase II
beta-catenin-TCF complex assembly
protein binding
RNA polymerase II C-terminal domain phosphoserine binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR15013374.hg.1 | ENST00000299601.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013374.hg.1 | ENST00000538386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR15013374.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR15013374.hg.1 | TC15001449.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | NR_109775.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | NR_109775.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | NR_109775.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009445.hg.1 | NR_109775.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000558949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000558949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | ENST00000558949.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | ENST00000557871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | NR_109775.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | NR_109775.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | NR_109775.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009445.hg.1 | NR_109775.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500169403.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1500169403.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500169403.hg.1 | TC1500009446.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1500169403.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1500169403.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000299601.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000315141.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000558949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000558949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | ENST00000558949.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_001286430.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1500009446.hg.1 | NM_138792.3 | Refseq |
